Learn More
Biomatik Corporation Recombinant Adiponectin (ADP), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU40108
The purity of Recombinant Adiponectin (ADP) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Adiponectin. Alternative Names: GBP28; ApM1; AdipoQ; Acrp30; ACDC; APM1; C1Q And Collagen Domain Containing; Adipocyte Complement-Related Protein of 30 KDa; Adipose Most Abundant Gene Transcript 1. UniProt: Q15848. Expression Region: Ala108-Asn244. Tag Info: two N-terminal Tags, His-tag and S-tag. Theoretical MW: 21.3 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5451