Learn More
Biomatik Corporation Recombinant Alpha-Fodrin (SPTAN1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU40213
The purity of Recombinant Alpha-Fodrin (SPTAN1) is greater than 95%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Alpha-Fodrin. Alternative Names: SPTAN1; NEAS; (ALPHA)II-SPECTRIN; Spectrin Alpha Non-Erythrocytic 1; Spectrin Alpha Chain,Brain; Spectrin, non-erythroid alpha subunit. UniProt: Q13813. Expression Region: Thr1573-Asp1742. Tag Info: N-terminal His-Tag. Theoretical MW: 20.4 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5460