Learn More
Biomatik Corporation Recombinant Annexin A1 (ANXA1), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU40291
The purity of Recombinant Annexin A1 (ANXA1) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Annexin A1. Alternative Names: ANX-A1; ANX1; LPC1; Lipocortin I; Chromobindin-9; Calpactin II; Phospholipase A2 inhibitory protein. UniProt: P04083. Expression Region: Met1-Asn346. Tag Info: two N-terminal Tags, His-tag and S-tag. Theoretical MW: 44.4 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5468