Learn More
Biomatik Corporation Recombinant Bruton'S Tyrosine Kinase (Btk), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU40528
The purity of Recombinant Bruton'S Tyrosine Kinase (Btk) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Bruton'S Tyrosine Kinase. Alternative Names: AGMX1; AT; ATK; BPK; IMD1; PSCTK1; XLA; Bruton Agammaglobulinemia Tyrosine Kinase; B-cell progenitor kinase. UniProt: Q06187. Expression Region: Glu396-Ser659. Tag Info: two N-terminal Tags, His-tag and GST-tag. Theoretical MW: 60.8 kDa. Storage Buffer: PBS, pH7.4, containing 0.01% SKL, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5489