Learn More
Biomatik Corporation Recombinant Carbohydrate Antigen 19-9 (CA19-9), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU40593
The purity of Recombinant Carbohydrate Antigen 19-9 (CA19-9) is greater than 95%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Carbohydrate Antigen 19-9. Alternative Names: sialylated Lewis (a)Antigen; Cancer Antigen 19-9. UniProt: P21217. Expression Region: Arg35-Thr361. Tag Info: N-terminal His-Tag. Theoretical MW: 39.7 kDa. Storage Buffer: PBS, pH7.4, containing 0.01% SKL, 1mM DTT, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5497