Learn More
Biomatik Corporation Recombinant Cholecystokinin (CCK), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU40778
The purity of Recombinant Cholecystokinin (CCK) is greater than 95%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Cholecystokinin. Alternative Names: CCK-PZ; Cholecystokinin-58 desnonopeptide. UniProt: P06307. Expression Region: Met1-Ser115. Tag Info: two N-terminal Tags, His-tag and GST-tag. Theoretical MW: 39.7 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5507