Learn More
Biomatik Corporation Recombinant Complement Component 3a (C3a), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU40945
The purity of Recombinant Complement Component 3a (C3a) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Mus musculus (Mouse). Gene Name: Complement Component 3a. UniProt: P01027. Expression Region: Ser671-Arg748. Tag Info: two N-terminal Tags, His-tag and GST-tag. Theoretical MW: 41.2 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5516