Learn More
Biomatik Corporation Recombinant Connective Tissue Growth Factor (CTGF), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU40982
The purity of Recombinant Connective Tissue Growth Factor (CTGF) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Mus musculus (Mouse). Gene Name: Connective Tissue Growth Factor. Alternative Names: CCN2; HCS24; IGFBP8; NOV2; Insulin-Like Growth Factor-Binding Protein 8; Hypertrophic Chondrocyte-Specific Protein 24; CCN Family Member 2. UniProt: P29268. Expression Region: Leu245-Asp346. Tag Info: N-terminal His-Tag. Theoretical MW: 13.2 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5522