Learn More
Biomatik Corporation Recombinant Connexin 40 (CX40), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU40993
The purity of Recombinant Connexin 40 (CX40) is greater than 98%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Rattus norvegicus (Rat). Gene Name: Connexin 40. Alternative Names: GJA5; GJ-A5; Gap Junction Protein Alpha 5. UniProt: P28234. Expression Region: Ala50-Glu303. Tag Info: N-terminal His-Tag. Theoretical MW: 31.3 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5524