Learn More
Biomatik Corporation Recombinant Corin (CRN), 100UG DFS Item
Manufacturer:  Biomatik Corporation RPU41005
The purity of Recombinant Corin (CRN) is greater than 92%. Source: E.coli. Bioactivity: Not tested. Endotoxin Level: <1.0EU per 1ug (determined by the LAL method). Applications: Positive Control; Immunogen; SDS-PAGE; WB. Species: Homo sapiens (Human). Gene Name: Corin. Alternative Names: ATC2; CRN; Lrp4; TMPRSS10; Atrial Natriuretic Peptide-Converting Enzyme, Serine Protease; Heart-specific serine proteinase ATC2; Pro-ANP-converting enzyme. UniProt: Q9Y5Q5. Expression Region: Arg134-Pro573. Tag Info: two N-terminal Tags, His-tag and T7-tag. Theoretical MW: 53.1 kDa. Storage Buffer: 20mM Tris, 150mM NaCl, pH8.0, containing 1mM EDTA, 1mM DTT, 0.01% SKL, 5% Trehalose and Proclin300. Storage: Short term: -20°C; Long term: -80°C. Minimize freeze and thaw cycles. Restriction: For Research Use Only.
Catalog No. 50-150-5525