Learn More
Alfa Aesar™ Cataxa/Rh(COD)/(S,S-Dipamp)
Manufacturer:  Alfa Aesar™ 4483206
Quantity 5g
 View more versions of this product
Catalog No. AA4483206
  Check Availability
Add to cart
Specifications
Description
  • This product is a Rhodium catalyst anchored on alumina via phosphotungstic acid
  • Used in enantioselective hydrogenation of prochiral olefinic bonds
Description & Specifications
Chemical Identifiers
MFCD08064220
UN3260
Cataxa/Rh (COD)/ (S,S-Dipamp)
Specifications
5g
MFCD08064220
Cataxa/Rh (COD)/ (S,S-Dipamp)
Powder
UN3260
Safety and Handling
SDS & Safety Documents
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
storageNote1 :
  • Packaged under argon