Alfa Aesar™ Catecholborane, 50% w/w in toluene (98% dry wt.)

Manufacturer: Alfa Aesar™ H3023414

Quantity 25g

 View more versions of this product

Catalog No. AAH3023414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 274-07-7
    Molecular Formula C6H5BO2
    Formula Weight 119.92
    MDL Number MFCD00005846
    UN Number UN2924
    Synonym 1,3,2-Benzodioxaborole
    Chemical Name or Material Catecholborane

    Specifications

    Formula Weight 119.92
    TSCA TSCA
    MDL Number MFCD00005846
    UN Number UN2924
    Health Hazard 2 GHS H Statement
    H225-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Concentration 50% w/w in toluene
    Quantity 25g
    Molecular Formula C6H5BO2
    CAS 274-07-7
    Synonym 1,3,2-Benzodioxaborole
    Health Hazard 1 Danger
    Chemical Name or Material Catecholborane
    Assay 98%, (dry wt.)

    Description

    Applications
    Employed in organic synthesis. Addition of catecholborane to vinylarenes in the presence of Wilkinson ft.s catalyst, gives Markovnikov hydroboration products exclusively, whereas the corresponding reaction with a-substituted vinylarenesgivesgood yields of vinylboronate. Hydroboration with catecholborane, followed by treatment with easily available reagents such as alkenyl sulfones or alkynyl phenyl sulfones in the presence of a radical initiator, represents an effective and simple one-pot procedure for direct vinylation, formylation, and cyanation. Catecholborane may be used as a stereoselective reducing agent when converting β-hydroxy ketones to syn 1,3-diols.

    Solubility
    Reacts with water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from water, moisture.

    GHS H Statement
    H225-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit