Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Catecholborane, 50% w/w in toluene (98% dry wt.)

Manufacturer:  Alfa Aesar™ H3023422

Quantity 100g

 View more versions of this product

Catalog No. AAH3023422


  Check Availability
Add to cart

Specifications

Description

Applications
Employed in organic synthesis. Addition of catecholborane to vinylarenes in the presence of Wilkinson ft.s catalyst, gives Markovnikov hydroboration products exclusively, whereas the corresponding reaction with a-substituted vinylarenesgivesgood yields of vinylboronate. Hydroboration with catecholborane, followed by treatment with easily available reagents such as alkenyl sulfones or alkynyl phenyl sulfones in the presence of a radical initiator, represents an effective and simple one-pot procedure for direct vinylation, formylation, and cyanation. Catecholborane may be used as a stereoselective reducing agent when converting β-hydroxy ketones to syn 1,3-diols.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from water, moisture.
Description & Specifications

Chemical Identifiers

274-07-7
119.92
UN2924
Catecholborane
C6H5BO2
MFCD00005846
1,3,2-Benzodioxaborole

Specifications

50% w/w in toluene
274-07-7
119.92
UN2924
Catecholborane
100g
C6H5BO2
MFCD00005846
1,3,2-Benzodioxaborole
98%, (dry wt.)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit