Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catenin alpha 1 Mouse anti-Human, PE/Cy5.5, Clone: 1G5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254533PECY55

Catalog No. N254533PEC5


Add to cart

Description

Description

Catenin alpha 1 Monoclonal antibody specifically detects Catenin alpha 1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Catenin alpha 1
1G5
Flow Cytometry
Conjugated, Purified
CTNNA1
Human alpha catenin fusion protein
Protein A or G purified
Cancer, Cellular Markers, Core ESC Like Genes, Cytoskeleton Markers, Phospho Specific, Stem Cell Markers
Primary
1495.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit