Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catenin alpha Mouse anti-Human, Clone: 1G5- Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

$378.35

Specifications

Antigen Catenin alpha
Clone 1G5
Host Species Mouse
Isotype IgG1
Gene Alias Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254533100 View Documents Novus Biologicals
NBP254533100UG
100 ug Each for $378.35
Add to cart
 
NB254533200 View Documents Novus Biologicals
NBP254533200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Catenin alpha 1 Monoclonal antibody specifically detects Catenin alpha 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications

Specifications

Catenin alpha
Mouse
Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Store at -20 to -80C. Avoid freeze-thaw cycles.
Monoclonal
1 mg/mL
1G5
IgG1
Human
RUO
Purified
Primary
PBS with No Preservative

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit