Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
BioVision IncSupplier Diversity Partner Cathepsin G Activity Colorimetric Assay Kit: Cathepsin-G Activity Assay Kit: Rapid & Convenient Colorimetric Assay to Measure Cathepsin-G Activity within 1-2 hrs. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K146100
Cathepsin G is an enzymatic protein belonging to the peptidase or protease families. The protein is found in azurophil granules of neutrophilic polymorphonuclear leukocytes. The encoded protease has a specificity similar to that of chymotrypsin C, and may participate in the killing and digestion of engulfed pathogens, and in connective tissue remodeling at sites of inflammation. In BioVision’s Cathepasin G Activity assay Kit, Cathepsin G will cleave the substrate and release the dye group, pNA (4-Nitroaniline), which can be detect at 405 nm. In presence of the Cathepsin G specific inhibitor, the cleavage will be stopped. The kit provides a rapid, simple, sensitive, and reliable test suitable for the activity of Cathepsin G. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9561