Learn More
BioVision IncSupplier Diversity Partner Cathepsin L Inhibitor Screening Kit (Fluorometric): Rapid & Convenient Fluorometric Assay to Screen/Study/Characterize Cathepsin L Inhibitors. Includes Inhibitor Control, FF-FMK. High-Throughput adaptable. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K161100
Cathepsin L (CTSL, EC 3.4.22.15) is a lysosomal cysteine protease that is implicated in protein degradation, arthritis, apoptosis, and cancer. Biovision’s Cathepsin L Inhibitor Screening Kit utilizes the ability of active Cathepsin L to cleave the synthetic AFC-based peptide substrate to release AFC, which can be easily quantified using a fluorometer or fluorescence microplate reader. In the presence of a Cathepsin L inhibitor, the cleavage of this substrate is reduced/abolished resulting in decrease or total loss of the AFC fluorescence. This simple and high-throughput adaptable assay kit can be used to screen/study/characterize potential inhibitors of Cathepsin L. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9576