Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathelicidin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific OSC00009W
Description
Glycerol (1:1) may be added for additional stability. Reconstitute with 150 µL of distilled water. OSC00009W can be used with blocking peptide GSC00002P.
Cathelicidin antimicrobial protein is an antimicrobial protein found in specific granules of polymorphonuclear leukocytes. Expressed in bone marrow and testis and neutrophils.Specifications
Cathelicidin | |
Polyclonal | |
CAMP | |
CAP-18, hCAP-18, antibacterial protein LL-37, CAMP, CRAMP, FALL39, hCAP18 | |
Rabbit | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
P49913 | |
CAMP | |
A synthetic peptide from human Cathelicidin antimicrobial peptide | |
RUO | |
820 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title