Learn More
Cathepsin 3 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0156-2K13, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212422AF594
Catalog No. NP212422A94
Add to cart
Description
Description
Cathepsin 3 Monoclonal antibody specifically detects Cathepsin 3 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Cathepsin 3
RM0156-2K13
Western Blot
1600000I23Rik
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Cts3
This antibody was produced from a hybridoma of a mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Cathepsin-3.
0.1 mL
Primary
117066