Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin 3 Antibody (RM0156-2K13) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
$204.00 - $542.50
Specifications
Antigen | Cathepsin 3 |
---|---|
Clone | RM0156-2K13 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
Cathepsin 3 Monoclonal specifically detects Cathepsin 3 in Mouse samples. It is validated for Western Blot.Specifications
Cathepsin 3 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
NP_081182 | |
117066 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0156-2K13 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
1600000I23Rik | |
Cts3 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title