Learn More
Alfa Aesar™ Cathepsin B Inhibitor IV
Manufacturer:  Alfa Aesar™ J64737MA
 View more versions of this product
Catalog No. AAJ64737MA
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
Specifications
Specifications
10mg
−30° to −10°C
A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
C19H31N3O6
CA-074 ME; (L-3-trans-(Propylcarbamyl)oxirane-2-carbonyl)-L-isoleucyl-L-proline methyl ester
Signal Transduction Reagent
White
Powder
147859-80-1
397.46g/mol
Soluble in DMSO