Learn More
Cathepsin F Mouse anti-Human, Alexa Fluor 647, Clone: MM0129-3A41, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212079AF647
Catalog No. NB212079647
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Cathepsin F Monoclonal antibody specifically detects Cathepsin F in Human samples. It is validated for Western Blot.
Specifications
Specifications
Cathepsin F
MM0129-3A41
Western Blot
Purified
CTSF
Human recombinant Cathepsin-F
Protein G purified
Immunology
Primary
8722
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
cathepsin F, CATSFEC 3.4.22.41, EC 3.4.22
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human