Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ Cathepsin G Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ Cathepsin G Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS1835R1
Description
Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.Host Species: Mouse
Clone: 19C3
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: N-terminal 124aa
Immunogen: Recombinant protein corresponding to 124aa from N-terminus of human cathepsin G
Molecular Weight: 29 kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Cathepsin G Ab-1 | |
Monoclonal | |
Unconjugated | |
Recombinant protein corresponding to 124aa from N terminus of human cathepsin G | |
RUO | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
19C3 | |
Mouse | |
1 mL | |
Apoptosis | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction