Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin K, Mouse anti-Human, DyLight 350, Clone: CTSK/2793, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279900UV

Catalog No. NB012161


Add to cart

Description

Description

Cathepsin K Monoclonal specifically detects Cathepsin K in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cathepsin K
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CTSK
Recombinant fragment of human Cathepsin K protein (around aa 163-274) (exact sequence is proprietary)
0.1 mL
Cancer
Human
IgG1 κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
CTSK/2793
Immunohistochemistry, Immunohistochemistry-Paraffin
cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit