Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin K Rabbit anti-Human, Mouse, Rat, Porcine, Chicken, Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Specifications
Antigen | Cathepsin K |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot, Immunohistochemistry, Immunoprecipitation |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP145460
|
Novus Biologicals
NBP145460 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Cathepsin K Polyclonal specifically detects Cathepsin K in Human, Mouse, Rat, Porcine, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Cathepsin K | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Polyclonal | |
Rabbit | |
Cancer | |
NP_113748 | |
1513 | |
Synthetic peptide surrounding amino acid 321 of rat cathepsin K (NP_113748). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Porcine, Chicken, Rabbit | |
cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD | |
CTSK | |
IgG | |
Affinity Purified | |
The antibody detects proform (43 kDa) and the mature form (29 kDa) of cathepsin K. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title