Learn More
Alfa Aesar™ Cathepsin K Substrate (fluorogenic)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J64082MB
 View more versions of this product
Catalog No. AAJ64082MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
White
Lyophilized
Fluorogenic
-30° to -10°C
A sensitive fluorogenic substrate for cathepsin K
578.65
25mg