Learn More
Cathepsin L1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP310283100UL
 View more versions of this product
Catalog No. NB125467
Add to cart
Description
Description
Cathepsin L1 Polyclonal antibody specifically detects Cathepsin L1 in Mouse samples. It is validated for Western Blot
Specifications
Specifications
Cathepsin L1
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
Primary
Mouse
Western Blot
Western Blot 1.0 ug/ml
Purified
The immunogen is a synthetic peptide directed towards the middle region of mouse Cathepsin L1 (NP_034114.1). Peptide sequence AMDASHPSLQFYSSGIYYEPNCSSKNLDHGVLLVGYGYEGTDSNKNKYWL
100 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal