Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin S Goat anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP145464

 View more versions of this product

Catalog No. NBP145464

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cathepsin S Polyclonal antibody specifically detects Cathepsin S in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

Cathepsin S
0.2 mg/mL
Western Blot, Immunohistochemistry, Immunoprecipitation
AAC37592
cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886
Rabbit
IgG
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
PBS (pH 7.2), 0.5% BSA, 30% Glycerol, 5mM EDTA with 0.03% Proclin
Affinity Purified
CTSS
Recombinant human Cathepsin S (AAC37592).
Immunogen affinity purified
RUO
Primary
1520
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only