Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin S Goat anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP1454640.05MG

Catalog No. NBP14546405


Add to cart

Description

Description

Cathepsin S Polyclonal antibody specifically detects Cathepsin S in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry,Immunoprecipitation.
Specifications

Specifications

Cathepsin S
0.20 mg/mL
Western Blot, Immunohistochemistry, Immunoprecipitation
AAC37592
CTSS
Recombinant human Cathepsin S (AAC37592).
0.05 mg
Primary
1520
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
PBS (pH 7.2), 0.5% BSA, 30% Glycerol, 5 mM EDTA with 0.03% Proclin
cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886
Goat
IgG
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit