Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cathepsin V Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112393
Description
Cathepsin V Polyclonal Antibody for Western Blot, IHC (P), ELISA
Cathepsin V (CTSU, CTSV, CATL2, cathepsin U, cathepsin L2) is a lysosomal cysteine proteinase that influences corneal physiology, and mediates degradation of invariant chain in human thymus. Cathepsin V is present in corneal epithelium, activated macrophages, and colorectal and breast carcinomas. A 1.8-kb mRNA to Cathepsin V is present in normal human thymus and testis. Cathepsin V is a member of the peptidase C1 family. Cysteine proteinases are synthesized as preproenzymes, which are processed to the corresponding proenzymes. The proenzymes are either targeted to the lysosome or continue along the cellular secretory route.
Specifications
Cathepsin V | |
Polyclonal | |
Unconjugated | |
CTSV | |
Cathepsin L2; cathepsin L2, preproprotein; cathepsin U; Cathepsin V; CATL2; CTSL2; CTSU; CTSV; PRO305; UNQ268; UNQ268/PRO305 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1515 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60911 | |
CTSV | |
Recombinant Human Cathepsin L2 protein (105-334AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction