Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CathepsinD Mouse anti-Human, Janelia Fluor 549, Clone: 4G2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP104278JF549

Catalog No. NBP14278549


Add to cart

Description

Description

CathepsinD Monoclonal antibody specifically detects CathepsinD in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CathepsinD
4G2
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
CTSD
Recombinant human Cathepsin D (21-412 aa) purified from E. coli (NP_001900).
Protein G purified
Cancer, Cellular Markers
Primary
1509
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10
Mouse
IgG2b Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Anti-human cathepsin D mAb, clone 4G2, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human cathepsin D protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit