Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CATIP Mouse anti-Human, Clone: OTI2E6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526146

Catalog No. PIMA526146


Add to cart

Description

Description

Plays a role in primary ciliogenesis by modulating actin polymerization.
Specifications

Specifications

CATIP
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q7Z7H3
CATIP, C2orf62
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
375307
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2E6
Unconjugated
CATIP
Liquid
CATIP
Full length human recombit protein of C2orf62 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit