Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CATPB, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1322598-09-3
C19H17ClF3NO3
399.794
QOSIJVVNNGXEKE-INIZCTEOSA-N
CATPB
53308747
(3S)-3-[[2-(3-chlorophenyl)acetyl]amino]-4-[4-(trifluoromethyl)phenyl]butanoic acid
C1=CC(=CC(=C1)Cl)CC(=O)NC(CC2=CC=C(C=C2)C(F)(F)F)CC(=O)O
This item is not returnable. View return policy
1322598-09-3
C19H17ClF3NO3
399.794
QOSIJVVNNGXEKE-INIZCTEOSA-N
CATPB
53308747
(3S)-3-[[2-(3-chlorophenyl)acetyl]amino]-4-[4-(trifluoromethyl)phenyl]butanoic acid
C1=CC(=CC(=C1)Cl)CC(=O)NC(CC2=CC=C(C=C2)C(F)(F)F)CC(=O)O

FFA2 antagonist/inverse agonist

FFA2 inverse agonist (pKi = 7.87). Selective for human over mouse FFA2. Increases forskolin-induced cAMP production and inhibits acetate-induced MAPK signaling in cells expressing human FFA2.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Linear Formula C19H17ClF3NO3
Solubility Information Soluble to 100mM in DMSO and to 50mM in ethanol
Formula Weight 399.79
Physical Form White Solid
Percent Purity >98%
Chemical Name or Material (S)-3-(2-(3-Chlorophenyl)acetamido)-4-(4-(trifluoromethyl)phenyl)butanoic acid

Recommended Storage : Store at Room Temperature