Description
- Fisher Science Education™ Bio-Fresh™ specimens are fixed in formaldehyde, rinsed and packaged in Bio-Fresh solution
- Fisher-Free™ specimens are 100% formaldehyde free — fixed and held in a nontoxic solution
- Plastic bags and ID tags included with each specimen
- Specimens are obtained using humane and socially acceptable procedures
- Unconditional warranty
Proper lab techniques, along with rinsing the specimen and using our Fisher Science Education Bio-Fresh spray, reduce odor effectively without compromising the specimen's useful life.
Order Info
INJECTION KEY
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue and hepatic portal veins yellow
Specifications
Cat | |
Double | |
45.72+ cm | |
1/Pk. |
Bio-Fresh™ | |
18+ in. | |
Plastic bags, ID tags |