Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CATSPER3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31101725UL
Description
CATSPER3 Polyclonal specifically detects CATSPER3 in Human samples. It is validated for Western Blot.Specifications
CATSPER3 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Ca(v)-like protein, CACRC, calcium channel repeat containing 1, cation channel sperm-associated protein 3, cation channel, sperm associated 3, CatSper3, MGC126741, One-repeat calcium channel-like protein, putative ion channel CatSper3 | |
The immunogen is a synthetic peptide directed towards the C terminal region of human CATSPER3 (NP_821138.1). | |
25 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
347732 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title