Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaV1.3 Mouse anti-Human, Mouse, Rat, Clone: S38-8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527715

Catalog No. PIMA527715


Add to cart

Description

Description

Voltage-sensitive calcium channels (VSCC) mediate the entry of calcium ions into excitable cells and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, gene expression, cell motility, cell division and cell death. The isoform alpha-1D gives rise to L-type calcium currents. Long-lasting (L-type) calcium channels belong to the high-voltage activated (HVA) group. They are blocked by dihydropyridines (DHP), phenylalkylamines, benzothiazepines, and by omega-agatoxin-IIIA (omega-Aga-IIIA). They are however insensitive to omega-conotoxin-GVIA (omega-CTx-GVIA) and omega-agatoxin-IVA (omega-Aga-IVA).
Specifications

Specifications

CaV1.3
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
P27732, Q01668, Q99246
alpha-1 polypeptide, CAC1D_HUMAN, CACH3, CACN4, CACNA 1D, Cacna1d, CACNL1A2, Calcium channel, Calcium channel L type alpha 1 polypeptide isoform 2, Calcium channel neuroendocrine/brain type alpha 1 subunit, Calcium channel voltage dependent L type alpha 1D subunit, CCHL1A2, isoform 2, L type, Voltage dependent L type calcium channel subunit alpha 1D, Voltage gated calcium channel alpha 1 subunit, Voltage gated calcium channel alpha subunit Cav1.3, Voltage gated calcium channel subunit alpha Cav1.3, Voltage-dependent L-type calcium channel subunit alpha-1D, Voltage-gated calcium channel subunit alpha Cav1.3
Mouse
IgG1
100 μg
-20°C
Primary
12289, 29716, 776
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
S38-8
Unconjugated
CACNA1D
Liquid
Cacna1d
Fusion protein amino acids 2025-2161 of rat Cav1.3
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit