Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caveolin-1 Antibody (7C8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280617
Description
Caveolin-1 Monoclonal specifically detects Caveolin-1 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Caveolin-1 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1:1000 - 1:4000, Flow Cytometry : 1 ug per million cells, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence : 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin : 1:100 - 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24758774) | |
CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Sheep | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
7C8 | |
Unconjugated | |
Tris-Glycine, 0.15 M NaCl | |
CAV1 | |
Intracellular membrane protein-containing vesicles (containing GLUT4) from rat adipocytes | |
0.1 mL | |
Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction | |
857 | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction