Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caveolin-1 Mouse anti-Human, Mouse, Rat, Sheep, PE/Atto594, Clone: 7C8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB100615PEATT594

Catalog No. N10615PE594

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Caveolin-1 Monoclonal antibody specifically detects Caveolin-1 in Human, Mouse, Rat, Sheep samples. It is validated for Flow Cytometry.
Specifications

Specifications

Caveolin-1
7C8
Flow Cytometry
Conjugated, Purified
CAV1
Intracellular membrane protein-containing vesicles (containing GLUT4) from rat adipocytes
Protein A purified
Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction
Primary
857.0
Human, Mouse, Rat, Sheep
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
This is specific for caveolin alpha and beta proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit