Learn More
Caveolin-1 (p Tyr14) Mouse anti-Human, Biotin, Clone: MM0680-4C29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236783B
Catalog No. NBP236783B
Add to cart
Description
Description
Caveolin-1 p Tyr14 Monoclonal antibody specifically detects Caveolin-1 p Tyr14 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Caveolin-1 (p Tyr14)
MM0680-4C29
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
CAV1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide Y14 site of human Caveolin-1.
Protein G purified
Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction
Primary
857
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human phosphor-Y14 Caveolin-1 in direct ELISA and WB.