Learn More
Caveolin-1 (p Tyr14) Mouse anti-Human, DyLight 755, Clone: MM0680-4C29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236783IR
Catalog No. NBP236783IR
Add to cart
Description
Description
Caveolin-1 p Tyr14 Monoclonal antibody specifically detects Caveolin-1 p Tyr14 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Caveolin-1 (p Tyr14)
MM0680-4C29
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
CAV1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide Y14 site of human Caveolin-1.
Protein G purified
Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction
Primary
857
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human phosphor-Y14 Caveolin-1 in direct ELISA and WB.