Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caveolin-2 Rabbit anti-Human, Clone: SY22-05, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267423
Description
Caveolin-2 Monoclonal antibody specifically detects Caveolin-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Caveolin-2 | |
SY22-05 | |
Unconjugated | |
Purified | |
CAV, caveolae protein, 20-kD, caveolin 2, caveolin 2 isoform a and b, caveolin-2, MGC12294 | |
CAV2 | |
Recombinant Protein | |
Monoclonal | |
Protein A purified | |
Caveolae Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
Human |
Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunoprecipitation,Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
858 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title