Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MedSchenker™ CavSwab™ Mid-Turbinate
SDP

The MedSchenker™ Mid-Turbinate CavSwab™ is recommended for pediatric use and has been engineered to provide a quicker and more comfortable testing process for both the medical professional as well as the patient.

Supplier:  MedSchenker™ AC3100

 View more versions of this product

Catalog No. 23-000-009


Add to cart

Description

Description

  • Marked nylon shaft with age recommended stop points, between 0 - 12+, for uncomplicated identification of required depth.
  • Removable stopper, the face of a bear, helps distract and put younger patients at ease.
  • Precision scalloped surface of ‘cavums’ provide a greater surface area to absorb and collect larger amounts of specimen.
  • Allium™ flock technology - nylon fibers adapt to the direction the swab is being pushed/pulled, resulting in less friction and an overall more pleasant experience for both the medical professional and the patient.
  • Notched shaft increases grip and dexterity while also reducing the amount of stress on the practitioner.
  • Electron beam sterilized and individually wrapped in pouches to ensure 100% sterility.
Specifications

Specifications

ISO 13485
Nylon Fiber
36 months
Flocked
Blue
100 Pk.
85 mm
Nylon Fiber
Nylon
Sterile
Sample Collection
120 mm
2000/Cs.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit