CB 13, Tocris

Potent dual CB1/CB2 agonist

Manufacturer: R&D Systems 2928/50

Quantity 50mg

 View more versions of this product

Catalog No. 29-285-0

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C26H24O2
    Formula Weight 368.47
    Synonym 1-Naphthalenyl[4-(pentyloxy)-1-naphthalenyl]methanone
    Chemical Name or Material CB 13

    Specifications

    Formula Weight 368.47
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C26H24O2
    CAS 432047-72-8
    Synonym 1-Naphthalenyl[4-(pentyloxy)-1-naphthalenyl]methanone
    Recommended Storage Store at Room Temperature
    Chemical Name or Material CB 13

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit