CB 65, Tocris

High affinity, selective CB2 agonist

Manufacturer: R&D Systems 2663/10

Quantity 10mg

 View more versions of this product

Catalog No. 26-631-0

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C22H28ClN3O3
    Formula Weight 417.93
    Synonym N-Cyclohexyl-7-chloro-1-[2-(4-morpholinyl)ethyl]quinolin-4(1H)-one-3-carboxamide
    Chemical Name or Material CB 65

    Specifications

    Formula Weight 417.93
    Quantity 10mg
    Assay Percent Range >99%
    Molecular Formula C22H28ClN3O3
    CAS 913534-05-1
    Recommended Storage Store at Room Temperature
    Synonym N-Cyclohexyl-7-chloro-1-[2-(4-morpholinyl)ethyl]quinolin-4(1H)-one-3-carboxamide
    Chemical Name or Material CB 65

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit