Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBFB Mouse anti-Human, Alexa Fluor 700, Clone: PCRP-CBFB-1E6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314059AF700
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
CBFB Monoclonal antibody specifically detects CBFB in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
CBFB | |
Monoclonal | |
Alexa Fluor 700 | |
CBF-beta, core-binding factor subunit beta, core-binding factor, beta subunit, PEA2-beta, PEBP2B, PEBP2-beta, polyomavirus enhancer binding protein 2, beta subunit, Polyomavirus enhancer-binding protein 2 beta subunit, SL3/AKV core-binding factor beta subunit, SL3-3 enhancer factor 1 beta subunit, SL3-3 enhancer factor 1 subunit beta | |
Recombinant full-length human CBFB protein (Uniprot: Q13951) | |
0.1 mL | |
DNA Replication Transcription Translation and Splicing | |
865 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-CBFB-1E6 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'