Learn More
CBFB Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP300328C
Catalog No. NB065586
Add to cart
Description
Description
CBFB Polyclonal antibody specifically detects CBFB in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
CBFB
DyLight 650
50mM Sodium Borate
CBF-beta, core-binding factor subunit beta, core-binding factor, beta subunit, PEA2-beta, PEBP2B, PEBP2-beta, polyomavirus enhancer binding protein 2, beta subunit, Polyomavirus enhancer-binding protein 2 beta subunit, SL3/AKV core-binding factor beta subunit, SL3-3 enhancer factor 1 beta subunit, SL3-3 enhancer factor 1 subunit beta
Produced in rabbits immunized with purified, recombinant CBFB (Q13951-1; Met1-Pro182).
Affinity purified
RUO
Primary
865
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human