Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
CBFB Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259243
Catalog No. NBP259243
Description
Description
CBFB Polyclonal antibody specifically detects CBFB in Human samples. It is validated for Chromatin Immunoprecipitation, ELISA.
Specifications
Specifications
CBFB
Unconjugated
Serum with 0.05% Sodium Azide
CBF-beta, core-binding factor subunit beta, core-binding factor, beta subunit, PEA2-beta, PEBP2B, PEBP2-beta, polyomavirus enhancer binding protein 2, beta subunit, Polyomavirus enhancer-binding protein 2 beta subunit, SL3/AKV core-binding factor beta subunit, SL3-3 enhancer factor 1 beta subunit, SL3-3 enhancer factor 1 subunit beta
Rabbit
IgG
100 ul
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
ChIP assay, ELISA
Chromatin Immunoprecipitation 4 ul, ELISA 1:500
Antiserum
CBFB
The exact immunogen is propietary information.
Whole Antiserum
RUO
Primary
865