CBiPES hydrochloride, Tocris™

Positive allosteric modulator of mGlu2

Manufacturer: R&D Systems 3949/50

Quantity 50mg

 View more versions of this product

Catalog No. 39-495-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C21H19N3O2S·HCl
    Formula Weight 413.92
    Synonym N-(4'-Cyano-[1, 1'-biphenyl]-3-yl-N-(3-pyridinylmethyl)-ethanesulfonamide hydrochloride
    Chemical Name or Material CBiPES hydrochloride

    Specifications

    Formula Weight 413.92
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C21H19N3O2S·HCl
    CAS 856702-40-4
    Synonym N-(4'-Cyano-[1, 1'-biphenyl]-3-yl-N-(3-pyridinylmethyl)-ethanesulfonamide hydrochloride
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material CBiPES hydrochloride

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit