Learn More
Azzota Corp CBL-0137 DFS Item
Manufacturer:  Azzota Corp C1118
CBL-0137 is an orally available curaxin-based agent targeting the Facilitates Chromatin Transcription (FACT) complex, with potential antineoplastic activity. Please request if need more quantities
Catalog No. 50-161-3096