Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cbl-c Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI4E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271756AF405

Catalog No. N271756A405


Add to cart

Description

Description

Cbl-c Monoclonal antibody specifically detects Cbl-c in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

Cbl-c
OTI4E3
Western Blot, Flow Cytometry
Affinity Purified, Conjugated
CBLC
Recombinant protein fragment corresponding to amino acids 244-474 of human Cbl-c (NP_036248) produced in HEK293T cells.
Immunogen affinity purified
Stem Cell Markers
Primary
23624.0
Flow Cytometry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit